missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Fluoro-4-formylbenzeneboronic acid pinacol ester, 96%

Brand:  Alfa Aesar™ H62784.03

Code : NEW

Additional Details : CAS Number : 503176-50-9 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15477836

  • £29.00 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

3-Fluoro-4-formylbenzeneboronic acid pinacol ester
250.07
3-Fluoro-4-formylphenylboronic acid pinacol ester
503176-50-9
MFCD16036135

Specifications

3-Fluoro-4-formylbenzeneboronic acid pinacol ester
503176-50-9
96%
MFCD16036135
1g
Solid
250.07
3-Fluoro-4-formylphenylboronic acid pinacol ester
Videos
SDS