missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-(4-morpholinyl)benzeneboronic acid pinacol ester, 95%, Thermo Scientific™

Catalog Number p-7040245
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
873431-46-0
C16H23BFNO3
307.172
MFCD22988989
YOJYRVSDQHWBGS-UHFFFAOYSA-N
4-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl morpholine, 3-fluoro-4-4-morpholinyl benzeneboronic acid pinacol ester
70975109
4-[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)N3CCOCC3)F
This item is not returnable. View return policy
873431-46-0
C16H23BFNO3
307.172
MFCD22988989
YOJYRVSDQHWBGS-UHFFFAOYSA-N
4-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl morpholine, 3-fluoro-4-4-morpholinyl benzeneboronic acid pinacol ester
70975109
4-[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)N3CCOCC3)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Boronate esters are air- and chromatography-stable and suitable for spectroscopic study. The Suzuki-Miyaura cross-coupling reaction can be used with boronate esters.

Notes
Store at room temperature. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 307.17
Percent Purity 95%
Chemical Name or Material 3-Fluoro-4-(4-morpholinyl)benzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'