missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-2-pyridinecarbonitrile, 95%, Thermo Scientific™

Catalog Number 10473953
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
97509-75-6
C6H3FN2
122.102
MFCD06797501
VZFPSCNTFBJZHB-UHFFFAOYSA-N
2-cyano-3-fluoropyridine, 3-fluoropicolinonitrile, 3-fluoro-2-pyridinecarbonitrile, 3-fluoro-2-cyanopyridine, 3-fluoro-2-pyridinenitrile, 3-fluoro-pyridine-2-carbonitrile, 2-pyridinecarbonitrile, 3-fluoro, fluorocyanopyridine, pubchem16434, acmc-209s8i
7060408
3-fluoropyridine-2-carbonitrile
C1=CC(=C(N=C1)C#N)F
This item is not returnable. View return policy
97509-75-6
C6H3FN2
122.102
MFCD06797501
VZFPSCNTFBJZHB-UHFFFAOYSA-N
2-cyano-3-fluoropyridine, 3-fluoropicolinonitrile, 3-fluoro-2-pyridinecarbonitrile, 3-fluoro-2-cyanopyridine, 3-fluoro-2-pyridinenitrile, 3-fluoro-pyridine-2-carbonitrile, 2-pyridinecarbonitrile, 3-fluoro, fluorocyanopyridine, pubchem16434, acmc-209s8i
7060408
3-fluoropyridine-2-carbonitrile
C1=CC(=C(N=C1)C#N)F
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 122.1
Percent Purity 95%
Chemical Name or Material 3-Fluoro-2-pyridinecarbonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'