missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-2-(methoxycarbonyl)benzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog Number p-7054325
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1293284-61-3
C14H18BFO4
280.102
KRBBXOCBQBADHY-UHFFFAOYSA-N
methyl 2-fluoro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 2-fluoro-6-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzoic acid methyl ester, benzoic acid, 2-fluoro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-, methyl ester, 3-fluoro-2-methoxycorbonyl benzeneboronic acid pinacol ester, 3-fluoro-2-methoxycorbonyl phenylboronic acid pinacol ester, 2-fluoro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzoic acid, methyl ester
67086691
methyl 2-fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
B1(OC(C(O1)(C)C)(C)C)C2=C(C(=CC=C2)F)C(=O)OC
This item is not returnable. View return policy
1293284-61-3
C14H18BFO4
280.102
KRBBXOCBQBADHY-UHFFFAOYSA-N
methyl 2-fluoro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 2-fluoro-6-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzoic acid methyl ester, benzoic acid, 2-fluoro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-, methyl ester, 3-fluoro-2-methoxycorbonyl benzeneboronic acid pinacol ester, 3-fluoro-2-methoxycorbonyl phenylboronic acid pinacol ester, 2-fluoro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzoic acid, methyl ester
67086691
methyl 2-fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
B1(OC(C(O1)(C)C)(C)C)C2=C(C(=CC=C2)F)C(=O)OC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 280.1
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 3-Fluoro-2-(methoxycarbonyl)benzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'