Alfa Aesar™ 3-Fluoro-2-iodotoluene, 95%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H63151.03

1GR 3-Fluoro-2-iodotoluene, 95% 1g

UNSPSC: 12352101

Code: NEW

Additional Details:
Additional Details: CAS Number: 883502-14-5 Weight: 0.00100kg


Quantity 1g

Product Code. 15429688

Quantity Price
1 £ 32.12 / 1g
Estimated Shipment date
from Supplier 08-02-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 3-Fluoro-2-iodotoluene
CAS 883502-14-5
Formula Weight 236.03
MDL Number MFCD04973438

Specification

Formula Weight 236.03
Chemical Name or Material 3-Fluoro-2-iodotoluene
Physical Form Solid
Assay 95%
MDL Number MFCD04973438
CAS 883502-14-5
Storage Note 1 -30° to -10°C
Sensitivity Light Sensitive
Quantity 1g
Safety and Handling

-30° to -10°C