missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Fluoro-2-(1-piperazinyl)benzonitrile, 95%

Brand:  Alfa Aesar™ H61759.03

Code : NEW

Additional Details : CAS Number : 1233026-65-7 Weight : 0.00100kg Transport : UN number : 3439 Chem class : 1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15458328

  • £406.10 / 1g


Estimated Shipment date
from Supplier 24-05-2018
Add to basket

Specifications

Chemical Identifiers

3-Fluoro-2-(1-piperazinyl)benzonitrile
1233026-65-7
205.23
UN3439

Specifications

3-Fluoro-2-(1-piperazinyl)benzonitrile
1233026-65-7
95%
UN3439
1g
Solid
205.23
Videos
SDS