missing translation for 'onlineSavingsMsg'
Learn More

3-Ethylphenol, 95%, Thermo Scientific Chemicals

Catalog Number p-7027030
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
620-17-7
C8H10O
122.17
MFCD00002311
HMNKTRSOROOSPP-UHFFFAOYSA-N
m-ethylphenol, phenol, 3-ethyl, phenol, m-ethyl, meta-ethylphenol, 1-ethyl-3-hydroxybenzene, 1-hydroxy-3-ethylbenzene, 3-ethyl-phenol, unii-0g9zk222jx, benzene, 1-ethyl-3-hydroxy, dsstox_cid_2480
12101
CHEBI:34332
3-ethylphenol
CCC1=CC=CC(O)=C1
This item is not returnable. View return policy
620-17-7
C8H10O
122.17
MFCD00002311
HMNKTRSOROOSPP-UHFFFAOYSA-N
m-ethylphenol, phenol, 3-ethyl, phenol, m-ethyl, meta-ethylphenol, 1-ethyl-3-hydroxybenzene, 1-hydroxy-3-ethylbenzene, 3-ethyl-phenol, unii-0g9zk222jx, benzene, 1-ethyl-3-hydroxy, dsstox_cid_2480
12101
CHEBI:34332
3-ethylphenol
CCC1=CC=CC(O)=C1

It is used as a photographic chemical intermediate and an intermediate for the cyan coupler of photographic paper. It has a wide application in fragrance industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a photographic chemical intermediate and an intermediate for the cyan coupler of photographic paper. It has a wide application in fragrance industry.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point -4°C
Density 1.028
Boiling Point 218°C
Flash Point 94°C (202°F)
Refractive Index 1.5335
Quantity 10 g
UN Number UN3145
Beilstein 1857002
Solubility Information Slightly soluble in water.
Formula Weight 122.17
Percent Purity 95%
Chemical Name or Material 3-Ethylphenol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'