missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-3-oxetanemethanol, 96%, ACROS Organics™

Brand:  Acros Organics 377700250

Code : 29

Additional Details : CAS Number : 3047-32-3 Weight : 0.07500kg


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10668034

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

3-Ethyl-3-oxetanemethanol
95.0
≥95%
C6H12O2
MFCD00190143
3047-32-3
100.0
96%
116.16

Specifications

3-Ethyl-3-oxetanemethanol
25g
95.0
100.0
Glass bottle
Liquid
96%
116.16
Clear colorless
3047-32-3
Authentic
1.019g/mL
1.4520 to 1.4540 (20°C, 589nm)
≥95%
C6H12O2
MFCD00190143
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3-Ethyl-3-oxetanemethanol

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS