missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxyphenylboronic acid, 97%, Thermo Scientific™

Catalog Number 10696650
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
90555-66-1
C8H11BO3
165.99
CHCWUTJYLUBETR-UHFFFAOYSA-N
3-ethoxyphenyl boronic acid, 3-ethoxybenzeneboronic acid, 3-ethoxyphenylboronicacid, 3-ethoxyphenyl boranediol, m-ethoxyphenylboronic acid, boronic acid, 3-ethoxyphenyl, pubchem1835, acmc-209r6q, 3-ethoxy-phenylboronic acid
2773920
(3-ethoxyphenyl)boronic acid
B(C1=CC(=CC=C1)OCC)(O)O
This item is not returnable. View return policy
90555-66-1
C8H11BO3
165.99
CHCWUTJYLUBETR-UHFFFAOYSA-N
3-ethoxyphenyl boronic acid, 3-ethoxybenzeneboronic acid, 3-ethoxyphenylboronicacid, 3-ethoxyphenyl boranediol, m-ethoxyphenylboronic acid, boronic acid, 3-ethoxyphenyl, pubchem1835, acmc-209r6q, 3-ethoxy-phenylboronic acid
2773920
(3-ethoxyphenyl)boronic acid
B(C1=CC(=CC=C1)OCC)(O)O
CAS Min % 96.0
CAS Max % 100.0
Melting Point 151.5°C to 152.2°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 165.99
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3-Ethoxyphenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Ethoxyphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'