missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxy-2,4-difluorobenzamide, 97%, Thermo Scientific™

Catalog Number p-7031586
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1017779-91-7
C9H9F2NO2
201.173
MFCD09258725
SQCQKFZIDMTPGR-UHFFFAOYSA-N
3-ethoxy-2,4-difluorobenzamide, jrd
46737642
3-ethoxy-2,4-difluorobenzamide
CCOC1=C(C=CC(=C1F)C(=O)N)F
This item is not returnable. View return policy
1017779-91-7
C9H9F2NO2
201.173
MFCD09258725
SQCQKFZIDMTPGR-UHFFFAOYSA-N
3-ethoxy-2,4-difluorobenzamide, jrd
46737642
3-ethoxy-2,4-difluorobenzamide
CCOC1=C(C=CC(=C1F)C(=O)N)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 100°C to 102°C
Quantity 1 g
Formula Weight 201.17
Percent Purity 97%
Chemical Name or Material 3-Ethoxy-2,4-difluorobenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'