missing translation for 'onlineSavingsMsg'
Learn More

3-Dimethylaminopropiophenone Hydrochloride 99%, ACROS Organics™

Brand:  Acros Organics 115891000

Code : 29

Additional Details : CAS Number : 879-72-1 Weight : 0.15000kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III


Quantity 100g
Packaging Glass bottle

 View more versions of this product

Product Code. 10723841

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

3-Dimethylaminopropiophenone hydrochloride
879-72-1
100.0
C11H15NO·HCl
213.71
99%
98.5
99%
C6H5COCH2CH2N(CH3)2·HCl
MFCD00012481

Specifications

3-Dimethylaminopropiophenone hydrochloride
100g
879-72-1
100.0
99%
C11H15NO·HCl
213.71
14, II, 38
152.0° to 155.0°C
99%
98.5
Glass bottle
99%
C6H5COCH2CH2N(CH3)2·HCl
MFCD00012481
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3-Dimethylaminopropiophenone hydrochloride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3

Hazard Statement
  • H301-Toxic if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

SDS