missing translation for 'onlineSavingsMsg'
Learn More

3-Dimethylaminobenzoic acid, 99%, Thermo Scientific Chemicals

Catalog Number p-115614
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
99-64-9
C9H11NO2
165.19
MFCD00002497
NEGFNJRAUMCZMY-UHFFFAOYSA-N
3-dimethylamino benzoic acid, benzoic acid, 3-dimethylamino, m-dimethylamino benzoic acid, m-dimethylaminobenzoic acid, n,n-dimethyl-m-aminobenzoic acid, 3-dimethylamino-benzoic acid, 3-n,n-dimethylaminobenzoic acid, benzoic acid, m-dimethylamino, m-dimethylamino-benzoic acid
66837
3-(dimethylamino)benzoic acid
CN(C)C1=CC=CC(=C1)C(=O)O
This item is not returnable. View return policy
99-64-9
C9H11NO2
165.19
MFCD00002497
NEGFNJRAUMCZMY-UHFFFAOYSA-N
3-dimethylamino benzoic acid, benzoic acid, 3-dimethylamino, m-dimethylamino benzoic acid, m-dimethylaminobenzoic acid, n,n-dimethyl-m-aminobenzoic acid, 3-dimethylamino-benzoic acid, 3-n,n-dimethylaminobenzoic acid, benzoic acid, m-dimethylamino, m-dimethylamino-benzoic acid
66837
3-(dimethylamino)benzoic acid
CN(C)C1=CC=CC(=C1)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 150.0°C to 153.0°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula (CH3)2NC6H4CO2H
Quantity 25 g
Beilstein 14, 392
Solubility Information Solubility in water: insoluble.
Formula Weight 165.19
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 3-Dimethylaminobenzoic acid, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'