missing translation for 'onlineSavingsMsg'
Learn More

3-(Dimethylamino)propionitrile, 98%, ACROS Organics™

Brand:  Acros Organics 215455000

Code : 29

Additional Details : CAS Number : 1738-25-6 Weight : 0.48500kg


Quantity 500mL
Packaging Glass bottle

 View more versions of this product

Product Code. 10022670

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

3-(Dimethylamino)propionitrile
1738-25-6
100.0
98%
(CH3)2NCH2CH2CN
MFCD00001957
98%
97.5
≥97.5%
C5H10N2
98.15

Specifications

3-(Dimethylamino)propionitrile
Clear colorless to yellow
98%
97.5
Authentic
1.4250 to 1.4270 (20°C, 589nm)
≥97.5%
C5H10N2
98.15
04, III, 1265
0.87g/mL
500mL
1738-25-6
100.0
Glass bottle
Liquid
98%
(CH3)2NCH2CH2CN
MFCD00001957
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3-(Dimethylamino)propionitrile

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS