missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 3-Diisopropylcarbamoyl-2-fluorobenzeneboronic acid, 97%
Brand: Alfa Aesar™ H56710.03
Code : NEW
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
3-Diisopropylcarbamoyl-2-fluorobenzeneboronic acid | |
267.11 | |
MFCD22682886 |
Specifications
3-Diisopropylcarbamoyl-2-fluorobenzeneboronic acid | |
97% | |
1g | |
MFCD22682886 |
Solid | |
White | |
267.11 |