missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Diethylamino-1-propyne, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
3-Diethylamino-1-propyne is used as functional leveling agent, brightener.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Diethylamino-1-propyne is used as functional leveling agent, brightener.
Solubility
Soluble in acid. Insoluble in water.
Notes
Incompatible with heat, light, carbondioxide, heavy metal powders. Air & Light Sensitive. Store in the dark. Store under dry inert gas.
3-Diethylamino-1-propyne is used as functional leveling agent, brightener.
Solubility
Soluble in acid. Insoluble in water.
Notes
Incompatible with heat, light, carbondioxide, heavy metal powders. Air & Light Sensitive. Store in the dark. Store under dry inert gas.
Specifications
Density | 0.804 |
Boiling Point | 119°C to 120°C |
Flash Point | 20°C (68°F) |
Odor | Amine-like |
Linear Formula | (CH3CH2)2NCH2C≡CH |
Refractive Index | 1.4305 |
Quantity | 5 g |
UN Number | UN2733 |
Beilstein | 1098468 |
Sensitivity | Air and light sensitive |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'