missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(Di-n-butylamino)-1-propyne, 98%

Brand:  Alfa Aesar™ L10844.03

Code : NEW

Additional Details : CAS Number : 6336-58-9 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 11363026

  • £21.00 / 1g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

3-(Di-n-butylamino)-1-propyne
C11H21N
MFCD00041673
N,N-Di-n-butylpropargylamine
6336-58-9
167.3
1752173

Specifications

3-(Di-n-butylamino)-1-propyne
87° to 89°C (19mmHg)
Amine-like
C11H21N
MFCD00041673
N,N-Di-n-butylpropargylamine
6336-58-9
1g
98%
167.3
1752173
Videos
SDS