missing translation for 'onlineSavingsMsg'
Learn More

3-Cyclopropyl-1H-pyrazole, 98%, Thermo Scientific Chemicals

Catalog Number 15401109
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
100114-57-6
C6H8N2
108.14
MFCD04970938,MFCD06739298
TXWDVWSJMDFNQY-UHFFFAOYSA-N
7017816
5-cyclopropyl-1H-pyrazole
C1CC1C1=CC=NN1
This item is not returnable. View return policy
100114-57-6
C6H8N2
108.14
MFCD04970938,MFCD06739298
TXWDVWSJMDFNQY-UHFFFAOYSA-N
7017816
5-cyclopropyl-1H-pyrazole
C1CC1C1=CC=NN1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Quantity 5 g
Beilstein 1561659
Synonym 3-cyclopropyl-1h-pyrazole, 5-cyclopropylpyrazole, 1h-pyrazole, 3-cyclopropyl, 3-cyclopropylpyrazole, 1h-pyrazole,3-cyclopropyl, 3-cyclopropyl-2h-pyrazole, 3-cyclopropyl-1h-pyrazole 97+%, acmc-2097ms, 1h-pyrazole, 5-cyclopropyl
Formula Weight 108.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Cyclopropyl-1H-pyrazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'