missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Cyclohexylhydantoin, 95%

Brand:  Alfa Aesar™ H63048.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15409508

  • £167.60 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

3-Cyclohexylhydantoin
182.22
MFCD12137796
3-Cyclohexyl-2,4-imidazolidinedione

Specifications

3-Cyclohexylhydantoin
1g
Solid
182.22
3-Cyclohexyl-2,4-imidazolidinedione
White
67518-23-4
95%
MFCD12137796
Videos
SDS