missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Cyclohexyl-1-phenyl-2-thiohydantoin, 95%

Brand:  Alfa Aesar™ H64482.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15461709

  • £597.84 / 5g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

3-Cyclohexyl-1-phenyl-2-thiohydantoin
274.38
MFCD28100492
3-Cyclohexyl-1-phenyl-2-thioxo-4-imidazolidinone

Specifications

3-Cyclohexyl-1-phenyl-2-thiohydantoin
Solid
274.38
3-Cyclohexyl-1-phenyl-2-thioxo-4-imidazolidinone
5g
95%
MFCD28100492
Videos
SDS