missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Cyclohexyl-1-methyl-2-thiohydantoin, 95%

Brand:  Alfa Aesar™ H63167.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15449708

  • £351.92 / 5g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

3-Cyclohexyl-1-methyl-2-thiohydantoin
212.31
MFCD02855391
3-Cyclohexyl-1-methyl-2-thioxo-4-imidazolidinone

Specifications

3-Cyclohexyl-1-methyl-2-thiohydantoin
Solid
212.31
3-Cyclohexyl-1-methyl-2-thioxo-4-imidazolidinone
5g
95%
MFCD02855391
Videos
SDS