missing translation for 'onlineSavingsMsg'
Learn More

3-Cyclohexenecarboxylic acid, 97%, ACROS Organics™

£22.23 - £147.86

Chemical Identifiers

CAS 4771-80-6
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.15
MDL Number MFCD00013781
InChI Key VUSWCWPCANWBFG-UHFFFAOYSA-N
Synonym 3-cyclohexene-1-carboxylic acid, 3-cyclohexenecarboxylic acid, cyclohex-3-enecarboxylic acid, 1,2,3,6-tetrahydrobenzoic acid, 1-cyclohexene-4-carboxylic acid, delta3-cyclohexenylcarboxylic acid, kyselina 1,2,5,6-tetrahydrobenzoova, 3-cyclohexene carboxylic acid, kyselina 1,2,5,6-tetrahydrobenzoova czech, .delta.3-cyclohexenecarboxylic acid
PubChem CID 20903
IUPAC Name cyclohex-3-ene-1-carboxylic acid
SMILES C1CC(CC=C1)C(=O)O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10157513
SDS View Documents
Acros Organics
294390010
1g Glass bottle
£22.23
1g
View alternative products
Save Up to 
»
In stock, ready to ship: 4 Please to purchase this item. Need a web account? Register with us today!
10033530
SDS View Documents
Acros Organics
294390100
10g Glass bottle
£147.86
10g
View alternative products
Save Up to 
»
Estimated Shipment
21-05-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

4771-80-6
126.15
VUSWCWPCANWBFG-UHFFFAOYSA-N
20903
C1CC(CC=C1)C(=O)O
C7H10O2
MFCD00013781
3-cyclohexene-1-carboxylic acid, 3-cyclohexenecarboxylic acid, cyclohex-3-enecarboxylic acid, 1,2,3,6-tetrahydrobenzoic acid, 1-cyclohexene-4-carboxylic acid, delta3-cyclohexenylcarboxylic acid, kyselina 1,2,5,6-tetrahydrobenzoova, 3-cyclohexene carboxylic acid, kyselina 1,2,5,6-tetrahydrobenzoova czech, .delta.3-cyclohexenecarboxylic acid
cyclohex-3-ene-1-carboxylic acid

Specifications

3-Cyclohexenecarboxylic acid
>112°C
Glass bottle
1.08
Blue-Gray to Brown-Yellow
1g
4771-80-6
100.0
C7H10O2
3-cyclohexene-1-carboxylic acid, 3-cyclohexenecarboxylic acid, cyclohex-3-enecarboxylic acid, 1,2,3,6-tetrahydrobenzoic acid, 1-cyclohexene-4-carboxylic acid, delta3-cyclohexenylcarboxylic acid, kyselina 1,2,5,6-tetrahydrobenzoova, 3-cyclohexene carboxylic acid, kyselina 1,2,5,6-tetrahydrobenzoova czech, .delta.3-cyclohexenecarboxylic acid
VUSWCWPCANWBFG-UHFFFAOYSA-N
cyclohex-3-ene-1-carboxylic acid
20903
Liquid
1.0800g/mL
Authentic
1.4800 to 1.4820
238.0°C
17.0°C
97%
96.0
96% min. (GC)
MFCD00013781
Solubility in water: insoluble. Other solubilities: soluble in methanol
C1CC(CC=C1)C(=O)O
126.15
126.15
97%
Safety and Handling

Safety and Handling

Product Identifier
  • 3-Cyclohexenecarboxylic acid

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS