missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanopyridine, 98%, Thermo Scientific Chemicals

Catalog Number p-95148
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
100-54-9
C6H4N2
104.11
MFCD00006372
GZPHSAQLYPIAIN-UHFFFAOYSA-N
3-cyanopyridine, nicotinonitrile, 3-pyridinecarbonitrile, nicotinic acid nitrile, 3-pyridinenitrile, 3-pyridylcarbonitrile, 3-azabenzonitrile, 3-cyjanopirydyna, nitryl kwasu nikotynowego, 3-cyjanopirydyna polish
79
CHEBI:86556
pyridine-3-carbonitrile
N#CC1=CC=CN=C1
This item is not returnable. View return policy
100-54-9
C6H4N2
104.11
MFCD00006372
GZPHSAQLYPIAIN-UHFFFAOYSA-N
3-cyanopyridine, nicotinonitrile, 3-pyridinecarbonitrile, nicotinic acid nitrile, 3-pyridinenitrile, 3-pyridylcarbonitrile, 3-azabenzonitrile, 3-cyjanopirydyna, nitryl kwasu nikotynowego, 3-cyjanopirydyna polish
79
CHEBI:86556
pyridine-3-carbonitrile
N#CC1=CC=CN=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 47.0°C to 51.0°C
Color Colorless or White to Yellow
Density 1.1590g/mL
Boiling Point 201.0°C
Flash Point 84°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (C5H4N)CN
Quantity 5 g
Beilstein 22, 41
Specific Gravity 1.159
Solubility Information Solubility in water: 140g/L (20°C)
Formula Weight 104.11
Percent Purity 98%
Physical Form Crystalline Mass, Crystals or Chunks
Chemical Name or Material 3-Cyanopyridine, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Cyanopyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'