missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanobenzaldehyde, 99%, Thermo Scientific Chemicals

Catalog Number p-95093
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
24964-64-5
MFCD00003344
HGZJJKZPPMFIBU-UHFFFAOYSA-N
3-cyanobenzaldehyde, m-cyanobenzaldehyde, benzonitrile, 3-formyl, m-formylbenzonitrile, m-cyanobenazldehyde, 3-cyano benzaldehyde, 3-formyl-benzonitrile, 3-formylbenzenecarbonitrile, 3cyano-benzaldehyde, m-cyano benzaldehyde
90670
3-formylbenzonitrile
C1=CC(=CC(=C1)C=O)C#N
This item is not returnable. View return policy
24964-64-5
MFCD00003344
HGZJJKZPPMFIBU-UHFFFAOYSA-N
3-cyanobenzaldehyde, m-cyanobenzaldehyde, benzonitrile, 3-formyl, m-formylbenzonitrile, m-cyanobenazldehyde, 3-cyano benzaldehyde, 3-formyl-benzonitrile, 3-formylbenzenecarbonitrile, 3cyano-benzaldehyde, m-cyano benzaldehyde
90670
3-formylbenzonitrile
C1=CC(=CC(=C1)C=O)C#N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 79°C to 81°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C8H5NO
Linear Formula NCC6H4CHO
Quantity 1 g
Beilstein 10, 671
Solubility Information Solubility in water: insoluble
Molecular Weight (g/mol) 131.13
Formula Weight 131.13
Percent Purity 99%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 3-Cyanobenzaldehyde, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Cyanobenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'