missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Cyano-5-(trifluoromethyl)benzeneboronic acid, 97%, Thermo Scientific™
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
3-Cyano-5-(trifluoromethyl)benzeneboronic acid is used as pharmaceutical intermediates.
Solubility
Slightly soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
3-Cyano-5-(trifluoromethyl)benzeneboronic acid is used as pharmaceutical intermediates.
Solubility
Slightly soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Quantity | 1 g |
Solubility Information | Slightly soluble in water. |
Formula Weight | 214.94 |
Percent Purity | 97% |
Physical Form | Solid |
Chemical Name or Material | 3-Cyano-5-(trifluoromethyl)benzeneboronic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'