missing translation for 'onlineSavingsMsg'
Learn More

3-Cyano-4-iodopyridine, 97%, Thermo Scientific Chemicals

Catalog Number p-4323309
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
490039-72-0
C6H3IN2
230.01
CJKHIIQONPNAIP-UHFFFAOYSA-N
3-cyano-4-iodopyridine, 4-iodonicotinonitrile, 3-pyridinecarbonitrile,4-iodo, acmc-20aibr, 4-iodo-nicotinonitrile, 4-iodo-3-pyridinecarbonitrile, 4-iodanylpyridine-3-carbonitrile
11138941
4-iodopyridine-3-carbonitrile
C1=CN=CC(=C1I)C#N
This item is not returnable. View return policy
490039-72-0
C6H3IN2
230.01
CJKHIIQONPNAIP-UHFFFAOYSA-N
3-cyano-4-iodopyridine, 4-iodonicotinonitrile, 3-pyridinecarbonitrile,4-iodo, acmc-20aibr, 4-iodo-nicotinonitrile, 4-iodo-3-pyridinecarbonitrile, 4-iodanylpyridine-3-carbonitrile
11138941
4-iodopyridine-3-carbonitrile
C1=CN=CC(=C1I)C#N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 114°C to 115°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 230.01
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3-Cyano-4-iodopyridine
compliance-icons
Product Identifier
  • 3-Cyano-4-iodopyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'