missing translation for 'onlineSavingsMsg'
Learn More

3-Cyano-2-pyridone, 98%, Thermo Scientific Chemicals

Catalog Number 15491359
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
20577-27-9
C6H4N2O
120.111
MFCD00661865
DYUMBFTYRJMAFK-UHFFFAOYSA-N
3-cyano-2-hydroxypyridine, 2-hydroxynicotinonitrile, 2-hydroxy-3-cyanopyridine, 2-hydroxypyridine-3-carbonitrile, 1,2-dihydro-2-oxopyridine-3-carbonitrile, 2-oxo-1,2-dihydropyridine-3-carbonitrile, 3-cyano-2 1h-pyridinone, 3-cyano-2-pyridone, 3-pyridinecarbonitrile, 1,2-dihydro-2-oxo, 2-oxo-1,2-dihydro-3-pyridinecarbonitrile
577543
2-oxo-1H-pyridine-3-carbonitrile
C1=CNC(=O)C(=C1)C#N
This item is not returnable. View return policy
20577-27-9
C6H4N2O
120.111
MFCD00661865
DYUMBFTYRJMAFK-UHFFFAOYSA-N
3-cyano-2-hydroxypyridine, 2-hydroxynicotinonitrile, 2-hydroxy-3-cyanopyridine, 2-hydroxypyridine-3-carbonitrile, 1,2-dihydro-2-oxopyridine-3-carbonitrile, 2-oxo-1,2-dihydropyridine-3-carbonitrile, 3-cyano-2 1h-pyridinone, 3-cyano-2-pyridone, 3-pyridinecarbonitrile, 1,2-dihydro-2-oxo, 2-oxo-1,2-dihydro-3-pyridinecarbonitrile
577543
2-oxo-1H-pyridine-3-carbonitrile
C1=CNC(=O)C(=C1)C#N

It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in dimethyl sulfoxide.

Notes
Store at -20°C. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point 224°C to 226°C
Quantity 5 g
UN Number UN3439
Beilstein 4661670
Solubility Information Soluble in dimethyl sulfoxide.
Formula Weight 120.11
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-Cyano-2-pyridone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'