missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Cyano-2-pyridone, 98%

Brand:  Alfa Aesar™ H64254.14

Code : NEW

Additional Details : CAS Number : 20577-27-9 Weight : 0.02500kg Transport : UN number : 3439 Chem class : 6.1 Pack group : III

Quantity 25g

 View more versions of this product

Product Code. 15401369

  • £76.75 / 25g

Estimated Shipment: 26-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Applications
It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in dimethyl sulfoxide.

Notes
Store at -20°C. Keep container tightly closed. Store away from oxidizing agent.
Specifications

Chemical Identifiers

20577-27-9
120.111
DYUMBFTYRJMAFK-UHFFFAOYSA-N
577543
C1=CNC(=O)C(=C1)C#N
C6H4N2O
MFCD00661865
3-cyano-2-hydroxypyridine, 2-hydroxynicotinonitrile, 2-hydroxy-3-cyanopyridine, 2-hydroxypyridine-3-carbonitrile, 1,2-dihydro-2-oxopyridine-3-carbonitrile, 2-oxo-1,2-dihydropyridine-3-carbonitrile, 3-cyano-2 1h-pyridinone, 3-cyano-2-pyridone, 3-pyridinecarbonitrile, 1,2-dihydro-2-oxo, 2-oxo-1,2-dihydro-3-pyridinecarbonitrile
2-oxo-1H-pyridine-3-carbonitrile

Specifications

3-Cyano-2-pyridone
25g
C6H4N2O
UN3439
3-cyano-2-hydroxypyridine, 2-hydroxynicotinonitrile, 2-hydroxy-3-cyanopyridine, 2-hydroxypyridine-3-carbonitrile, 1,2-dihydro-2-oxopyridine-3-carbonitrile, 2-oxo-1,2-dihydropyridine-3-carbonitrile, 3-cyano-2 1h-pyridinone, 3-cyano-2-pyridone, 3-pyridinecarbonitrile, 1,2-dihydro-2-oxo, 2-oxo-1,2-dihydro-3-pyridinecarbonitrile
DYUMBFTYRJMAFK-UHFFFAOYSA-N
2-oxo-1H-pyridine-3-carbonitrile
577543
Solid
224°C to 226°C
20577-27-9
MFCD00661865
4661670
Soluble in dimethyl sulfoxide.
C1=CNC(=O)C(=C1)C#N
120.111
120.11
98%
Videos
SDS Documents
Certificates