missing translation for 'onlineSavingsMsg'
Learn More

3-Cyano-2-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog Number 15477046
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1218791-15-1
C13H15BFNO2
247.076
MFCD11506358
BDIHSSFYJYXWRV-UHFFFAOYSA-N
2-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 3-cyano-2-fluorophenylboronic acid pinacol ester, 3-cyano-2-fluorophenylboronic acid,pinacol ester, 3-cyano-2-fluorophenylboronic acid, pinacol ester, 3-cyano-2-fluoro benzene boronic acid, pinacol ester, 2-fluoro-3-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, benzonitrile, 2-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
53217116
2-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
B1(OC(C(O1)(C)C)(C)C)C2=C(C(=CC=C2)C#N)F
This item is not returnable. View return policy
1218791-15-1
C13H15BFNO2
247.076
MFCD11506358
BDIHSSFYJYXWRV-UHFFFAOYSA-N
2-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 3-cyano-2-fluorophenylboronic acid pinacol ester, 3-cyano-2-fluorophenylboronic acid,pinacol ester, 3-cyano-2-fluorophenylboronic acid, pinacol ester, 3-cyano-2-fluoro benzene boronic acid, pinacol ester, 2-fluoro-3-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, benzonitrile, 2-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
53217116
2-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
B1(OC(C(O1)(C)C)(C)C)C2=C(C(=CC=C2)C#N)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 247.07
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 3-Cyano-2-fluorobenzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'