Learn More
3-CPMT, Tocris Bioscience™
Dopamine uptake inhibitor
Brand:  Tocris (Bio-Techne) 091750
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13488153
  • £266.00 / 50mg
Estimated Shipment
02-07-2018
Add to basket
Description
Description
This cocaine analog binds with high affinity to the dopamine transporter (Ki = 30 nM) and blocks re-uptake of dopamine. However, it is a weak psychomotor stimulant and does not substitute for cocaine in addicted rats.
Specifications
Chemical Identifiers
3-CPMT
C21H24ClNO·HCl
378.34
3α-[(4-Chlorophenyl)phenylmethoxy] tropane hydrochloride
Specifications
3-CPMT
14008-79-8
C21H24ClNO·HCl
3α-[(4-Chlorophenyl)phenylmethoxy] tropane hydrochloride
50mg
>99%
378.34
Videos