missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chlorotoluene, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
250 mL
500 mL
2.5 L
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -48°C |
CAS Min % | 96.0 |
Color | Colorless to Yellow |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Linear Formula | CH3C6H4Cl |
Beilstein | 05, 291 |
Merck Index | 15, 2175 |
Solubility Information | Solubility in water: slightly soluble |
Formula Weight | 126.59 |
Show More |
Safety and Handling
Product Identifier
- 3-Chlorotoluene
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Flammable liquid Category 3
- H411-Toxic to aquatic life with long lasting effects.
- H332-Harmful if inhaled.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'