missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropyridine-4-carboxaldehyde, 98%, Thermo Scientific Chemicals

Catalog Number 15402228
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
72990-37-5
C6H4ClNO
141.554
MFCD06200712
ZVGDKOQPJCOCLI-UHFFFAOYSA-N
3-chloroisonicotinaldehyde, 3-chloropyridine-4-carboxaldehyde, 3-chloro-4-pyridinecarboxaldehyde, 3-chloro-4-pyridinecarbaldehyde, 3-chloro-pyridine-4-carbaldehyde, 3-chloro-4-formylpyridine, 4-pyridinecarboxaldehyde, 3-chloro, acmc-1bgrb, ksc377a6f
2762995
3-chloropyridine-4-carbaldehyde
C1=CN=CC(=C1C=O)Cl
This item is not returnable. View return policy
72990-37-5
C6H4ClNO
141.554
MFCD06200712
ZVGDKOQPJCOCLI-UHFFFAOYSA-N
3-chloroisonicotinaldehyde, 3-chloropyridine-4-carboxaldehyde, 3-chloro-4-pyridinecarboxaldehyde, 3-chloro-4-pyridinecarbaldehyde, 3-chloro-pyridine-4-carbaldehyde, 3-chloro-4-formylpyridine, 4-pyridinecarboxaldehyde, 3-chloro, acmc-1bgrb, ksc377a6f
2762995
3-chloropyridine-4-carbaldehyde
C1=CN=CC(=C1C=O)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 58°C to 62°C
Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 141.56
Percent Purity 98%
Chemical Name or Material 3-Chloropyridine-4-carboxaldehyde

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'