missing translation for 'onlineSavingsMsg'
Learn More

(3-chloropropyl)trimethoxysilane, 98+%, Thermo Scientific Chemicals

Catalog Number p-3757273
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
2530-87-2
C6H15ClO3Si
198.72
MFCD00000997
OXYZDRAJMHGSMW-UHFFFAOYSA-N
3-chloropropyl trimethoxysilane, silane, 3-chloropropyl trimethoxy, 3-chloropropyltrimethyoxysilane, sila-ace s 620, cps-m, silane 3-chloropropyl tris methoxy, 3-trimethoxysilyl propyl chloride, unii-t21bnl1s7f, cptmo, 3-chloropropyl trimethoxysilan
62449
3-chloropropyl(trimethoxy)silane
CO[Si](CCCCl)(OC)OC
This item is not returnable. View return policy
2530-87-2
C6H15ClO3Si
198.72
MFCD00000997
OXYZDRAJMHGSMW-UHFFFAOYSA-N
3-chloropropyl trimethoxysilane, silane, 3-chloropropyl trimethoxy, 3-chloropropyltrimethyoxysilane, sila-ace s 620, cps-m, silane 3-chloropropyl tris methoxy, 3-trimethoxysilyl propyl chloride, unii-t21bnl1s7f, cptmo, 3-chloropropyl trimethoxysilan
62449
3-chloropropyl(trimethoxy)silane
CO[Si](CCCCl)(OC)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0700g/mL
Boiling Point 195°C to 196°C (750.0 mmHg)
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula (CH3O)3Si(CH2)3Cl
Refractive Index 1.4190 to 1.421
Quantity 25 mL
Specific Gravity 1.07
Solubility Information Solubility in water: soluble
Formula Weight 198.72
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material (3-Chloropropyl)trimethoxysilane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'