missing translation for 'onlineSavingsMsg'
Learn More

(3-Chloropropyl)triethoxysilane, 97+%, Thermo Scientific Chemicals

Catalog Number p-3593193
Click to view available options
Quantity:
5 g
25 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
5089-70-3
C9H21ClO3Si
240.8
MFCD00018985
KSCAZPYHLGGNPZ-UHFFFAOYSA-N
3-chloropropyl triethoxysilane, 3-chloropropyl triethoxy silane, silane, 3-chloropropyl triethoxy, unii-x7rb20518m, triethoxy gamma-chloropropyl silane, gamma-chloropropyltriethoxysilane, triethoxy .gamma.-chloropropyl silane, chloropropyl triethoxysilane, dynasylan cpteo
78771
3-chloropropyl(triethoxy)silane
CCO[Si](CCCCl)(OCC)OCC
This item is not returnable. View return policy
5089-70-3
C9H21ClO3Si
240.8
MFCD00018985
KSCAZPYHLGGNPZ-UHFFFAOYSA-N
3-chloropropyl triethoxysilane, 3-chloropropyl triethoxy silane, silane, 3-chloropropyl triethoxy, unii-x7rb20518m, triethoxy gamma-chloropropyl silane, gamma-chloropropyltriethoxysilane, triethoxy .gamma.-chloropropyl silane, chloropropyl triethoxysilane, dynasylan cpteo
78771
3-chloropropyl(triethoxy)silane
CCO[Si](CCCCl)(OCC)OCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.0
CAS Max % 100.0
Color Yellow
Density 1.0040g/mL
Boiling Point 230.0°C
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Linear Formula Cl(CH2)3Si(OCH2CH3)
Refractive Index 1.4190 to 1.4210 (20°C, 589nm)
Quantity 5 g
Specific Gravity 1.004
Solubility Information Solubility in water: insoluble
Formula Weight 240.8
Percent Purity 97+%
Physical Form Liquid
Chemical Name or Material (3-Chloropropyl)triethoxysilane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'