missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropropyl isocyanate, 97%, Thermo Scientific Chemicals

Catalog Number p-7046713
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
13010-19-0
C4H6ClNO
119.55
MFCD00010384
RQAVSDINDRNIKL-UHFFFAOYSA-N
3-chloropropyl isocyanate, 3-chloropropylisocyanate, propane, 1-chloro-3-isocyanato, 1-chloro-3-isocyanato-propane, chloropropylisocyanate, chloropropyl isocyanate, 3-chloropropanisocyanate, chloro propyl isocyanate, acmc-20an5q, 3-chloropropyl-isocyanate
166752
1-chloro-3-isocyanatopropane
ClCCCN=C=O
This item is not returnable. View return policy
13010-19-0
C4H6ClNO
119.55
MFCD00010384
RQAVSDINDRNIKL-UHFFFAOYSA-N
3-chloropropyl isocyanate, 3-chloropropylisocyanate, propane, 1-chloro-3-isocyanato, 1-chloro-3-isocyanato-propane, chloropropylisocyanate, chloropropyl isocyanate, 3-chloropropanisocyanate, chloro propyl isocyanate, acmc-20an5q, 3-chloropropyl-isocyanate
166752
1-chloro-3-isocyanatopropane
ClCCCN=C=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.165
Boiling Point 152°C to 154°C
Flash Point 61°C (141°F)
Odor Pungent
Refractive Index 1.452
Quantity 1 g
UN Number UN2206
Beilstein 1098789
Sensitivity Moisture sensitive
Formula Weight 119.55
Percent Purity 97%
Chemical Name or Material 3-Chloropropyl isocyanate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'