missing translation for 'onlineSavingsMsg'
Learn More

3'-Chloropropiophenone, 98%, Thermo Scientific Chemicals

Catalog Number p-91440
Click to view available options
Quantity:
25 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
34841-35-5
C9H9ClO
168.62
MFCD00009925
PQWGFUFROKIJBO-UHFFFAOYSA-N
3'-chloropropiophenone, 1-3-chlorophenyl propan-1-one, m-chloropropiophenone, 3-chlorophenyl ethyl ketone, 1-propanone, 1-3-chlorophenyl, 1-3-chloro-phenyl-propan-1-one, m-chloro-propiophenone, pubchem3083, 3'-chloro-propiophenone, 3/'-chloropropiophenone
587128
1-(3-chlorophenyl)propan-1-one
CCC(=O)C1=CC(=CC=C1)Cl
This item is not returnable. View return policy
34841-35-5
C9H9ClO
168.62
MFCD00009925
PQWGFUFROKIJBO-UHFFFAOYSA-N
3'-chloropropiophenone, 1-3-chlorophenyl propan-1-one, m-chloropropiophenone, 3-chlorophenyl ethyl ketone, 1-propanone, 1-3-chlorophenyl, 1-3-chloro-phenyl-propan-1-one, m-chloro-propiophenone, pubchem3083, 3'-chloro-propiophenone, 3/'-chloropropiophenone
587128
1-(3-chlorophenyl)propan-1-one
CCC(=O)C1=CC(=CC=C1)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 43°C to 47°C
Color White to Yellow
Boiling Point 124°C (14.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula ClC6H4COC2H5
Quantity 25 g
Beilstein 07, 1028
Solubility Information Solubility in water: soluble in hot water. Other solubilities: soluble in hot toluene and alcohol
Formula Weight 168.62
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material 3'-Chloropropiophenone, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3'-Chloropropiophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'