missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropropionic Acid 98%, ACROS Organics™

Brand:  Acros Organics 158151000

Code : 29

Additional Details : CAS Number : 107-94-8 Weight : 0.15000kg Transport : UN number : 3261 Chem class : 8 Pack group : II

Quantity 100g

 View more versions of this product

Product Code. 10476810

  • £13.70 / 100g


EU Stock 5
Add to basket

Specifications

Chemical Identifiers

3-Chloropropionic acid
107-94-8
100.0
98%
ClCH2CH2CO2H
MFCD00002764
98%
97.5
98%
C3H5ClO2
108.52

Specifications

3-Chloropropionic acid
203.0° to 205.0°C
37.0° to 41.0°C
98%
97.5
Authentic
Crystalline Low Melting Mass
98%
ClCH2CH2CO2H
MFCD00002764
15, 2161
125°C
White to beige
100g
107-94-8
100.0
Glass bottle
98%
C3H5ClO2
108.52
02, 249
Videos
SDS