missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropropanesulfonyl chloride, 98+%, Thermo Scientific Chemicals

Catalog Number p-7080899
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1633-82-5
C3H6Cl2O2S
177.039
MFCD00007463
GPKDGVXBXQTHRY-UHFFFAOYSA-N
3-chloropropanesulfonyl chloride, 3-chloro-1-propanesulfonyl chloride, 3-chloropropanesulfonylchloride, 3-chloropropylsulfonyl chloride, 3-chloropropanesulphonyl chloride, 1-propanesulfonyl chloride, 3-chloro, propanesulfonyl chloride, chloro, 3-chloro-propane-1-sulfonyl chloride, gamma-chloropropanesulfonyl chloride, 3-chloropropylsulfonylchloride
15410
3-chloropropane-1-sulfonyl chloride
C(CS(=O)(=O)Cl)CCl
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1633-82-5
C3H6Cl2O2S
177.039
MFCD00007463
GPKDGVXBXQTHRY-UHFFFAOYSA-N
3-chloropropanesulfonyl chloride, 3-chloro-1-propanesulfonyl chloride, 3-chloropropanesulfonylchloride, 3-chloropropylsulfonyl chloride, 3-chloropropanesulphonyl chloride, 1-propanesulfonyl chloride, 3-chloro, propanesulfonyl chloride, chloro, 3-chloro-propane-1-sulfonyl chloride, gamma-chloropropanesulfonyl chloride, 3-chloropropylsulfonylchloride
15410
3-chloropropane-1-sulfonyl chloride
C(CS(=O)(=O)Cl)CCl

3-Chloropropanesulfonyl Chloride is an intermediate used in the preparation of many enzymatic inhibitors. It is also used in the generation and trapping of the derived sulfene with imines and glyoxylates in the presence of chinchona alkaloids provide chiral sultams and sultones. It is mainly used for the synthesis of pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloropropanesulfonyl Chloride is an intermediate used in the preparation of many enzymatic inhibitors. It is also used in the generation and trapping of the derived sulfene with imines and glyoxylates in the presence of chinchona alkaloids provide chiral sultams and sultones. It is mainly used for the synthesis of pharmaceutical intermediates.

Solubility
Soluble in dichloromethane, ethyl Acetate and hexane.

Notes
Store at -20°C. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Moisture sensitive. Keep away from alcohols, strong bases, oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 1.456
Boiling Point 70°C (0.5 mmHg)
Flash Point >110°C (230°F)
Linear Formula Cl(CH2)3SO2Cl
Refractive Index 1.49
Quantity 10 g
UN Number UN3265
Beilstein 1754119
Sensitivity Moisture sensitive
Solubility Information Soluble in dichloromethane,ethyl Acetate and hexane.
Formula Weight 177.05
Percent Purity ≥98%
Chemical Name or Material 3-Chloropropanesulfonyl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'