missing translation for 'onlineSavingsMsg'
Learn More

3-chlorophenylacetylene, 97%, ACROS Organics™

Brand:  Acros Organics 433130010

Code : 29

Additional Details : CAS Number : 766-83-6 Weight : 0.05100kg


Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10712942

  • £35.30 / 1g


Estimated Shipment
14-05-2018
Add to basket


Specifications

Chemical Identifiers

3-chlorophenylacetylene
96.0
≥96.0%
C8H5Cl
MFCD03839984
766-83-6
100.0
97%
136.58
3-Chloro-1-ethynylbenzene

Specifications

3-chlorophenylacetylene
1g
96.0
Authentic
Liquid
97%
136.58
3-Chloro-1-ethynylbenzene
Clear colorless to yellow
766-83-6
100.0
Glass bottle
≥96.0%
C8H5Cl
MFCD03839984
Videos
SDS