missing translation for 'onlineSavingsMsg'
Learn More

3-chlorophenylacetylene, 97%, Thermo Scientific Chemicals

Catalog Number 10657770
106.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
766-83-6
C8H5Cl
136.58
MFCD03839984
GRBJPHPMYOUMJV-UHFFFAOYSA-N
3-chlorophenylacetylene, 3-chlorophenyl acetylene, 3'-chlorophenyl acetylene, 3-chloro-1-ethynylbenzene, benzene, 1-chloro-3-ethynyl, m-chlorophenylacetylene, 1-chloro-3-ethynyl-benzene, 3'-chlorophenylacetylene, 3-chloro phenyl acetylene
123086
1-chloro-3-ethynylbenzene
C#CC1=CC(=CC=C1)Cl
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
766-83-6
C8H5Cl
136.58
MFCD03839984
GRBJPHPMYOUMJV-UHFFFAOYSA-N
3-chlorophenylacetylene, 3-chlorophenyl acetylene, 3'-chlorophenyl acetylene, 3-chloro-1-ethynylbenzene, benzene, 1-chloro-3-ethynyl, m-chlorophenylacetylene, 1-chloro-3-ethynyl-benzene, 3'-chlorophenylacetylene, 3-chloro phenyl acetylene
123086
1-chloro-3-ethynylbenzene
C#CC1=CC(=CC=C1)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
Color Colorless to Yellow
CAS Max % 100.0
Boiling Point 178°C to 179°C
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Assay Percent Range 96% min. (GC)
Formula Weight 136.58
Percent Purity 97%
Chemical Name or Material 3-chlorophenylacetylene
Show More Show Less
compliance-icons
Product Identifier
  • 3-Chlorophenylacetylene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'