missing translation for 'onlineSavingsMsg'
Learn More

3-chlorophenylacetic acid, 99+%, ACROS Organics™

POA

Chemical Identifiers

CAS 1878-65-5
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00004332
InChI Key WFPMUFXQDKMVCO-UHFFFAOYSA-N
Synonym 3-chlorophenylacetic acid, m-chlorophenylacetic acid, 2-3-chlorophenyl acetic acid, 3-chlorobenzeneacetic acid, benzeneacetic acid, 3-chloro, m-chlorophenyl acetic acid, 3-chlorophenyl acetic acid, 3-chlorophenylaceticacid, acetic acid, m-chlorophenyl
PubChem CID 15879
IUPAC Name 2-(3-chlorophenyl)acetic acid
SMILES OC(=O)CC1=CC=CC(Cl)=C1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10010820
SDS View Documents
Acros Organics
109840100
10g Glass bottle
POA
10g
View alternative products
Save Up to 
»
N/A Please to purchase this item. Need a web account? Register with us today!
This item cannot be ordered online because the price is subject to change. Please contact our Product Support team on 01509 555500 for assistance.
10107573
SDS View Documents
Acros Organics
109840500
50g Glass bottle
POA
50g
View alternative products
Save Up to 
»
N/A Please to purchase this item. Need a web account? Register with us today!
This item cannot be ordered online because the price is subject to change. Please contact our Product Support team on 01509 555500 for assistance.
Description

Description

Specifications

Chemical Identifiers

1878-65-5
170.59
WFPMUFXQDKMVCO-UHFFFAOYSA-N
15879
OC(=O)CC1=CC=CC(Cl)=C1
C8H7ClO2
MFCD00004332
3-chlorophenylacetic acid, m-chlorophenylacetic acid, 2-3-chlorophenyl acetic acid, 3-chlorobenzeneacetic acid, benzeneacetic acid, 3-chloro, m-chlorophenyl acetic acid, 3-chlorophenyl acetic acid, 3-chlorophenylaceticacid, acetic acid, m-chlorophenyl
2-(3-chlorophenyl)acetic acid

Specifications

3-Chlorophenylacetic acid
Glass bottle
77°C to 80°C
1878-65-5
100.0
C8H7ClO2
MFCD00004332
3-chlorophenylacetic acid, m-chlorophenylacetic acid, 2-3-chlorophenyl acetic acid, 3-chlorobenzeneacetic acid, benzeneacetic acid, 3-chloro, m-chlorophenyl acetic acid, 3-chlorophenyl acetic acid, 3-chlorophenylaceticacid, acetic acid, m-chlorophenyl
WFPMUFXQDKMVCO-UHFFFAOYSA-N
2-(3-chlorophenyl)acetic acid
15879
Shiny Flakes and Chunks
Authentic
White
10g
99.0
99+%
ClC6H4CH2CO2H
09, II, 306
Solubility in water: slightly soluble. Other solubilities: soluble in methanol
OC(=O)CC1=CC=CC(Cl)=C1
170.59
170.6
99+%
Safety and Handling

Safety and Handling

Product Identifier
  • 3-Chlorophenylacetic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Product Certifications