missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorophenethylalcohol, 98%, ACROS Organics™

Brand:  Acros Organics 185960250

Code : 29

Additional Details : CAS Number : 5182-44-5 Weight : 0.08000kg

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10150262

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

3-Chlorophenethylalcohol
5182-44-5
100.0
98%
ClC6H4CH2CH2OH
MFCD00002892
98%
97.5
98%
C8H9ClO
156.61

Specifications

3-Chlorophenethylalcohol
>110°C
135.0° to 137.0°C (13.0 mmHg)
25g
5182-44-5
100.0
Glass bottle
Liquid
98%
ClC6H4CH2CH2OH
MFCD00002892
1.1810g/mL
1.181
Clear colorless to very slightly yellow
98%
97.5
Authentic
1.5475 to 1.5495
98%
C8H9ClO
156.61
Videos
SDS