missing translation for 'onlineSavingsMsg'
Learn More

3-(chloromethyl)heptane, 90%, ACROS Organics™

Brand:  Acros Organics 344980250

Code : 29

Additional Details : CAS Number : 123-04-6 Weight : 0.07205kg Transport : UN number : 1993 Chem class : 1 Pack group : III


Quantity 25mL
Packaging Glass bottle

 View more versions of this product

Product Code. 10506251

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

3-(Chloromethyl)heptane
88.0
90%
C8H17Cl
148.68
2-Ethylhexylchloride, Isooctylchloride
123-04-6
100.0
90%
H3C(CH2)3CH(CH2Cl)CH2CH3
MFCD00018940

Specifications

3-(Chloromethyl)heptane
88.0
Authentic
1.4320 to 1.4360
− 70.0°C
90%
90%
25mL
C8H17Cl
148.68
2-Ethylhexylchloride, Isooctylchloride
123-04-6
100.0
Glass bottle
0.8820g/mL
Liquid
Clear yellow
166.0° to 168.0°C
60°C
H3C(CH2)3CH(CH2Cl)CH2CH3
MFCD00018940
Videos
SDS