missing translation for 'onlineSavingsMsg'
Learn More

3-(chloromethyl)benzoyl chloride, 98%, ACROS Organics™

Brand:  Acros Organics 314430250

Code : 29

Additional Details : CAS Number : 63024-77-1 Weight : 0.07500kg Transport : UN number : 3265 Chem class : 1a Pack group : III

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10637164

  • £129.40 / 25g


EU Stock 1
Add to basket

Specifications

Chemical Identifiers

3-(chloromethyl)benzoyl chloride
97.5
≥97.5%
C8H6Cl2O
189.04
63024-77-1
100.0
98%
ClCHC6H4COCl
MFCD00000682

Specifications

3-(chloromethyl)benzoyl chloride
Clear colorless to light yellow
Authentic
97.5
Glass bottle
Liquid
98%
ClCHC6H4COCl
MFCD00000682
1.33g/mL
25g
63024-77-1
100.0
1.5740 to 1.5760 (20°C, 589nm)
≥97.5%
C8H6Cl2O
189.04
Videos
SDS