missing translation for 'onlineSavingsMsg'
Learn More

3-(chloromethyl)benzamide, 97%, Thermo Scientific™

Catalog Number 10714583
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
135654-16-9
C8H8ClNO
169.608
MFCD00175948
OOYATKQQGVPYQR-UHFFFAOYSA-N
3-chloromethyl benzamide, benzamide,3-chloromethyl, benzamide, 3-chloromethyl-9ci, 3-chloromethyl-benzamide, 3-carbamoylbenzyl chloride, acmc-20a41t, 3-aminocarbonyl benzyl chloride, 3-carbamoylbenzyl chloride, 3-aminocarbonyl benzyl chloride
737145
3-(chloromethyl)benzamide
C1=CC(=CC(=C1)CCl)C(=O)N
This item is not returnable. View return policy
135654-16-9
C8H8ClNO
169.608
MFCD00175948
OOYATKQQGVPYQR-UHFFFAOYSA-N
3-chloromethyl benzamide, benzamide,3-chloromethyl, benzamide, 3-chloromethyl-9ci, 3-chloromethyl-benzamide, 3-carbamoylbenzyl chloride, acmc-20a41t, 3-aminocarbonyl benzyl chloride, 3-carbamoylbenzyl chloride, 3-aminocarbonyl benzyl chloride
737145
3-(chloromethyl)benzamide
C1=CC(=CC(=C1)CCl)C(=O)N
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CH2ClC6H4C(O)NH2
Quantity 250 mg
Formula Weight 169.61
Percent Purity 97%
Chemical Name or Material 3-(chloromethyl)benzamide
compliance-icons
Product Identifier
  • 3-(Chloromethyl)benzamide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'