missing translation for 'onlineSavingsMsg'
Learn More

3-(Chloromethyl)-1,2,4-oxadiazole, Technical Grade, Thermo Scientific™

Catalog Number 10283322
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
51791-12-9
C3H3ClN2O
118.52
MFCD00052097
YSNKGJCEHOJIDK-UHFFFAOYSA-N
2735765
3-(chloromethyl)-1,2,4-oxadiazole
ClCC1=NOC=N1
This item is not returnable. View return policy
51791-12-9
C3H3ClN2O
118.52
MFCD00052097
YSNKGJCEHOJIDK-UHFFFAOYSA-N
2735765
3-(chloromethyl)-1,2,4-oxadiazole
ClCC1=NOC=N1
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 3-chloromethyl-1,2,4-oxadiazole, 1,2,4-oxadiazole,3-chloromethyl, chloromethyl 1,2,4-oxadiazole, 1,2,4-oxadiazole, 3-chloromethyl
Formula Weight 118.52
Percent Purity Tech.
Grade Technical
Chemical Name or Material 3-(Chloromethyl)-1, 2, 4-oxadiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'