missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Chloromethyl)-1,2,4-oxadiazole, Technical Grade, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Synonym | 3-chloromethyl-1,2,4-oxadiazole, 1,2,4-oxadiazole,3-chloromethyl, chloromethyl 1,2,4-oxadiazole, 1,2,4-oxadiazole, 3-chloromethyl |
Formula Weight | 118.52 |
Percent Purity | Tech. |
Grade | Technical |
Chemical Name or Material | 3-(Chloromethyl)-1, 2, 4-oxadiazole |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'