missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzylamine, 97%, Thermo Scientific Chemicals

Catalog Number p-7021659
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
4152-90-3
C7H8ClN
141.598
MFCD00040752
BJFPYGGTDAYECS-UHFFFAOYSA-N
3-chlorobenzylamine, 3-chlorophenyl methanamine, benzenemethanamine, 3-chloro, 1-3-chlorophenyl methanamine, m-chlorobenzylamine, 3-chloro-benzylamine, m-chlorobenzyl amine, 3-chlorophenyl methylamine, chembl12957, c2a
77802
(3-chlorophenyl)methanamine
C1=CC(=CC(=C1)Cl)CN
This item is not returnable. View return policy
4152-90-3
C7H8ClN
141.598
MFCD00040752
BJFPYGGTDAYECS-UHFFFAOYSA-N
3-chlorobenzylamine, 3-chlorophenyl methanamine, benzenemethanamine, 3-chloro, 1-3-chlorophenyl methanamine, m-chlorobenzylamine, 3-chloro-benzylamine, m-chlorobenzyl amine, 3-chlorophenyl methylamine, chembl12957, c2a
77802
(3-chlorophenyl)methanamine
C1=CC(=CC(=C1)Cl)CN

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.159
Boiling Point 226°C
Flash Point 98°C (208°F)
Odor Amine-like
Refractive Index 1.559
Quantity 10 g
UN Number UN2735
Beilstein 774509
Sensitivity Air Sensitive
Formula Weight 141.6
Percent Purity ≥97%
Chemical Name or Material 3-Chlorobenzylamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'