missing translation for 'onlineSavingsMsg'
Learn More

3-chlorobenzonitrile, 99%, Thermo Scientific Chemicals

Catalog Number p-3757207
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
766-84-7
C7H4ClN
137.57
MFCD00001798
WBUOVKBZJOIOAE-UHFFFAOYSA-N
m-chlorobenzonitrile, benzonitrile, 3-chloro, m-chloro benzonitrile, m-cyanochlorobenzene, benzonitrile, m-chloro, 3-chlorobenzenecarbonitrile, 5-chlorobenzonitrile, 3-chloro benzonitrile, 3-chloro-benzonitrile, pubchem3624
13015
3-chlorobenzonitrile
ClC1=CC=CC(=C1)C#N
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
766-84-7
C7H4ClN
137.57
MFCD00001798
WBUOVKBZJOIOAE-UHFFFAOYSA-N
m-chlorobenzonitrile, benzonitrile, 3-chloro, m-chloro benzonitrile, m-cyanochlorobenzene, benzonitrile, m-chloro, 3-chlorobenzenecarbonitrile, 5-chlorobenzonitrile, 3-chloro benzonitrile, 3-chloro-benzonitrile, pubchem3624
13015
3-chlorobenzonitrile
ClC1=CC=CC(=C1)C#N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 38°C to 42°C
CAS Min % 98.5
Color Colorless to Yellow
CAS Max % 100.0
Boiling Point 94°C (11.0 mmHg)
Quantity 10 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Assay Percent Range 98.5% min. (GC)
Linear Formula ClC6H4CN
Beilstein 09, 339
Solubility Information Solubility in water: insoluble
Formula Weight 137.57
Percent Purity 99%
Chemical Name or Material 3-Chlorobenzonitrile
Show More Show Less
compliance-icons
Product Identifier
  • 3-Chlorobenzonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'