missing translation for 'onlineSavingsMsg'
Learn More

3-Chloroaniline, 99%, Thermo Scientific Chemicals

Catalog Number p-7081850
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
108-42-9
C6H6ClN
127.57
MFCD00007765
PNPCRKVUWYDDST-UHFFFAOYSA-N
m-chloroaniline, 3-chlorobenzenamine, benzenamine, 3-chloro, m-chloraniline, 3-chlorophenylamine, orange gc base, m-chlorophenylamine, m-aminochlorobenzene, m-chloroaminobenzene, aniline, m-chloro
7932
3-chloroaniline
NC1=CC=CC(Cl)=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
108-42-9
C6H6ClN
127.57
MFCD00007765
PNPCRKVUWYDDST-UHFFFAOYSA-N
m-chloroaniline, 3-chlorobenzenamine, benzenamine, 3-chloro, m-chloraniline, 3-chlorophenylamine, orange gc base, m-chlorophenylamine, m-aminochlorobenzene, m-chloroaminobenzene, aniline, m-chloro
7932
3-chloroaniline
NC1=CC=CC(Cl)=C1

3-Chloroaniline was the only aniline compound to increase plasmaALT/GPT activity at 48 h. It is employed in the bioaugmentation of activated sludge.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloroaniline was the only aniline compound to increase plasmaALT/GPT activity at 48 h. It is employed in the bioaugmentation of activated sludge.

Solubility
Soluble in water. [6.8 g/L (20°C)].

Notes
Store in cool place. Keep away from oxidizing agents. Incompatible with bases.
TRUSTED_SUSTAINABILITY
Melting Point -10°C
Density 1.21
Boiling Point 230°C
Quantity 100 g
Odor Characteristic
Refractive Index 1.594
UN Number UN2019
Beilstein 605969
Merck Index 14,2118
Solubility Information Soluble in water. [6.8g/L (20°C)].
Formula Weight 127.57
Percent Purity 99%
Chemical Name or Material 3-Chloroaniline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'