missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Chloro-N-methylbenzamide, 97%

Brand:  Alfa Aesar™ H56565.03

Code : NEW

Additional Details : CAS Number : 18370-10-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15403068

  • £315.00 / 1g

Estimated Shipment: 23-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Notes
Incompatible with oxidizing agents. Store at room temperature.
Specifications

Chemical Identifiers

18370-10-0
169.608
WJJLGPQRUCWHKZ-UHFFFAOYSA-N
4150792
CNC(=O)C1=CC(=CC=C1)Cl
C8H8ClNO
MFCD01214034
3-chloro-n-methyl-benzamide, benzamide, 3-chloro-n-methyl, 3-chlorophenyl-n-methylcarboxamide, 5-chloro-n-methylbenzamide, n-methyl-3-chlorobenzamide
3-chloro-N-methylbenzamide

Specifications

3-Chloro-N-methylbenzamide
18370-10-0
MFCD01214034
WJJLGPQRUCWHKZ-UHFFFAOYSA-N
3-chloro-N-methylbenzamide
4150792
97%
1g
C8H8ClNO
3-chloro-n-methyl-benzamide, benzamide, 3-chloro-n-methyl, 3-chlorophenyl-n-methylcarboxamide, 5-chloro-n-methylbenzamide, n-methyl-3-chlorobenzamide
CNC(=O)C1=CC(=CC=C1)Cl
169.608
169.61
Videos
SDS Documents
Certificates